Assay Details
Gene Symbol: GRM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015130351.2 | XP_014985837.1 | 2760 | 87 | ||
XM_015130352.2 | XP_014985838.1 | 2760 | 87 | |||
XM_028844388.1 | XP_028700221.1 | 2761 | 87 | |||
GenBank mRNA | JU327979.1 | - | 651 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
