Assay Details
| Gene Symbol: GPR137B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126959.2 | XP_014982445.1 | 1265 | 84 | ||
| GenBank mRNA | JU336150.1 | - | 1089 | 84 | ||
| JV592243.1 | - | 1110 | 84 | |||
| JV722264.1 | - | 1071 | 84 | |||
More Information
Set Membership: |
|

GPR137B
Genomic Map