Assay Details
| Gene Symbol: KLHL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260584.1 | NP_001247513.1 | 1272 | 66 | ||
| XM_028843228.1 | XP_028699061.1 | 1367 | 66 | |||
| XM_028843229.1 | XP_028699062.1 | 1447 | 66 | |||
| XM_028843231.1 | XP_028699064.1 | 1461 | 66 | |||
| XM_028843232.1 | XP_028699065.1 | 1314 | 66 | |||
| XM_028843233.1 | XP_028699066.1 | 1356 | 66 | |||
| XM_028843234.1 | XP_028699067.1 | 1489 | 66 | |||
| GenBank mRNA | JU335116.1 | - | 1230 | 66 | ||
| JU335117.1 | - | 1230 | 66 | |||
| JU472060.1 | - | 1260 | 66 | |||
| JU472061.1 | - | 1260 | 66 | |||
| JV044208.1 | - | 1272 | 66 | |||
More Information
Set Membership: |
|

KLHL24
Genomic Map