Assay Details
| Gene Symbol: PAX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107687.4 | XP_001107687.2 | 967 | 103 | ||
| XM_028831147.1 | XP_028686980.1 | 963 | 103 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAX3| Gene Symbol: PAX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107687.4 | XP_001107687.2 | 967 | 103 | ||
| XM_028831147.1 | XP_028686980.1 | 963 | 103 | |||
Set Membership: |
|