Assay Details
| Gene Symbol: CDK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261340.1 | NP_001248269.1 | 228 | 73 | ||
| XM_015120714.2 | XP_014976200.1 | 739 | 73 | |||
| XM_015120715.1 | XP_014976201.1 | 389 | 73 | |||
| XM_015120716.2 | XP_014976202.1 | 95 | 73 | |||
| XM_028837144.1 | XP_028692977.1 | 739 | 73 | |||
| GenBank mRNA | JU332455.1 | - | 208 | 73 | ||
| JU473922.1 | - | 205 | 73 | |||
| JV045870.1 | - | 228 | 73 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015120716.2, XM_028837144.1 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CDK8
Genomic Map