Assay Details
| Gene Symbol: GMPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135594.2 | XP_014991080.1 | 668 | 68 | ||
| GenBank mRNA | JU334057.1 | - | 504 | 68 | ||
| JV046791.1 | - | 537 | 68 | |||
| JV648608.1 | - | 548 | 68 | |||
More Information
Set Membership: |
|

GMPR
Genomic Map