Assay Details
| Gene Symbol: UBE2E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193865.1 | NP_001180794.1 | 513 | 74 | ||
| XM_028843152.1 | XP_028698985.1 | 771 | 74 | |||
| XM_028843153.1 | XP_028698986.1 | 695 | 74 | |||
| GenBank mRNA | JU320368.1 | - | 563 | 74 | ||
| JU477054.1 | - | 513 | 74 | |||
| JV048142.1 | - | 526 | 74 | |||
| JV660451.1 | - | 518 | 74 | |||
| JV704429.1 | - | 600 | 74 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

UBE2E2
Genomic Map