Assay Details
| Gene Symbol: UNC93B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001104582.4 | XP_001104582.1 | 1010 | 55 | ||
| GenBank mRNA | JU494289.1 | - | 867 | 55 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UNC93B1| Gene Symbol: UNC93B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001104582.4 | XP_001104582.1 | 1010 | 55 | ||
| GenBank mRNA | JU494289.1 | - | 867 | 55 | ||
Set Membership: |
|