Assay Details
| Gene Symbol: MGLL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015130091.2 | XP_014985577.1 | 1080 | 65 | ||
| XM_015130092.2 | XP_014985578.1 | 495 | 65 | |||
| XM_015130093.2 | XP_014985579.1 | 755 | 65 | |||
| GenBank mRNA | JU321055.1 | - | 699 | 65 | ||
| JU321616.1 | - | 271 | 65 | |||
| JV604907.1 | - | 272 | 65 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MGLL
Genomic Map