Assay Details
Gene Symbol: TLCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260969.1 | NP_001247898.1 | 273 | 126 | ||
GenBank mRNA | JU333081.1 | - | 310 | 126 | ||
JU333082.1 | - | 310 | 126 | |||
JU333083.1 | - | 310 | 126 | |||
JU333084.1 | - | 310 | 126 | |||
JU333085.1 | - | 310 | 126 | |||
JV046261.1 | - | 273 | 126 | |||
JV643225.1 | - | 399 | 126 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
