Assay Details
| Gene Symbol: STS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001088752.4 | XP_001088752.2 | 743 | 105 | ||
| XM_015126901.2 | XP_014982387.1 | 779 | 105 | |||
| XM_015126902.2 | XP_014982388.1 | 662 | 105 | |||
| XM_028842249.1 | XP_028698082.1 | 779 | 105 | |||
| GenBank mRNA | JU330794.1 | - | 205 | 105 | ||
| JV715084.1 | - | 390 | 105 | |||
More Information
Set Membership: |
|

STS
Genomic Map