Assay Details
| Gene Symbol: APEX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261365.1 | NP_001248294.1 | 651 | 86 | ||
| GenBank mRNA | JU323181.1 | - | 642 | 86 | ||
| JU473317.1 | - | 653 | 86 | |||
| JV045278.1 | - | 651 | 86 | |||

APEX2
Genomic Map