Assay Details
| Gene Symbol: VCP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117511.2 | XP_014972997.1 | 2883 | 64 | ||
| XM_015117512.2 | XP_014972998.1 | 2473 | 64 | |||
| GenBank mRNA | JU320667.1 | - | 2308 | 64 | ||
| JU471040.1 | - | 2371 | 64 | |||
| JV044850.1 | - | 2364 | 64 | |||
| JV636831.1 | - | 2383 | 64 | |||
More Information
Set Membership: |
|

VCP
Genomic Map