Assay Details
| Gene Symbol: PGRMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194918.1 | NP_001181847.1 | 595 | 67 | ||
| GenBank mRNA | JU321574.1 | - | 578 | 67 | ||
| JU472080.1 | - | 534 | 67 | |||
| JV045603.1 | - | 545 | 67 | |||
| JV528454.1 | - | 602 | 67 | |||
| JV703299.1 | - | 596 | 67 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PGRMC1
Genomic Map