Assay Details
| Gene Symbol: WARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105363.4 | XP_001105363.2 | 697 | 114 | ||
| XM_001105786.4 | XP_001105786.2 | 1339 | 114 | |||
| XM_001105926.4 | XP_001105926.2 | 1151 | 114 | |||
| GenBank mRNA | JU336252.1 | - | 926 | 114 | ||
| JU477086.1 | - | 866 | 114 | |||
| JV048166.1 | - | 822 | 114 | |||
| JV556837.1 | - | 863 | 114 | |||
| JV636359.1 | - | 863 | 114 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

WARS1
Genomic Map