Assay Details
| Gene Symbol: YY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144480.2 | XP_014999966.1 | 1757 | 100 | ||
| GenBank mRNA | JU328424.1 | - | 339 | 100 | ||
| JU472038.1 | - | 895 | 100 | |||
| JV045322.1 | - | 888 | 100 | |||
| JV636361.1 | - | 966 | 100 | |||
More Information
Set Membership: |
|

YY1
Genomic Map