Assay Details
| Gene Symbol: MGST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151208.2 | XP_015006694.1 | 407 | 87 | ||
| GenBank mRNA | JU325570.1 | - | 113 | 87 | ||
| JV045789.1 | - | 225 | 87 | |||
| JV469005.1 | - | 290 | 87 | |||
| JV684416.1 | - | 281 | 87 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MGST1
Genomic Map