Assay Details
Gene Symbol: ACBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091471.4 | XP_001091471.1 | 1149 | 72 | ||
GenBank mRNA | JU326664.1 | - | 885 | 72 | ||
JV658933.1 | - | 1137 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
