Assay Details
| Gene Symbol: CARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266326.1 | NP_001253255.1 | 807 | 71 | ||
| XM_015113364.2 | XP_014968850.1 | 951 | 71 | |||
| XM_015113365.2 | XP_014968851.1 | 640 | 71 | |||
| XM_015113367.2 | XP_014968853.1 | 702 | 71 | |||
| XM_015113368.2 | XP_014968854.1 | 687 | 71 | |||
| XM_015113369.2 | XP_014968855.1 | 937 | 71 | |||
| XM_015113370.2 | XP_014968856.1 | 749 | 71 | |||
| GenBank mRNA | JU326766.1 | - | 614 | 71 | ||
| JU477414.1 | - | 807 | 71 | |||
| JV044168.1 | - | 575 | 71 | |||
More Information
Set Membership: |
|

CARS1
Genomic Map