Assay Details
| Gene Symbol: HNF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194548.3 | NP_001181477.1 | 562 | 95 | ||
| XM_001112182.4 | XP_001112182.2 | 535 | 95 | |||
| XM_028835662.1 | XP_028691495.1 | 1021 | 95 | |||
More Information
Set Membership: |
|

HNF1B
Genomic Map