Assay Details
| Gene Symbol: TIMM21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193707.2 | NP_001180636.1 | 586 | 90 | ||
| GenBank mRNA | JU323421.1 | - | 591 | 90 | ||
| JU473644.1 | - | 589 | 90 | |||
| JV043484.1 | - | 582 | 90 | |||
| JV638695.1 | - | 598 | 90 | |||
| JV704345.1 | - | 586 | 90 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TIMM21
Genomic Map