Assay Details
| Gene Symbol: PSMB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260845.2 | NP_001247774.1 | 210 | 72 | ||
| GenBank mRNA | JU335537.1 | - | 198 | 72 | ||
| JU476465.1 | - | 198 | 72 | |||
| JV047706.1 | - | 210 | 72 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PSMB5
Genomic Map