Assay Details
| Gene Symbol: OTUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113917.2 | XP_014969403.1 | 767 | 66 | ||
| GenBank mRNA | JU328225.1 | - | 119 | 66 | ||
| JU476317.1 | - | 369 | 66 | |||
| JV047586.1 | - | 159 | 66 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

OTUB1
Genomic Map