Assay Details
| Gene Symbol: SRGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261094.2 | NP_001248023.1 | 114 | 62 | ||
| GenBank mRNA | JU335343.1 | - | 145 | 62 | ||
| JU476295.1 | - | 165 | 62 | |||
| JV047576.1 | - | 116 | 62 | |||
| JV718293.1 | - | 154 | 62 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SRGN
Genomic Map