Assay Details
| Gene Symbol: UBL4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266701.1 | NP_001253630.1 | 167 | 104 | ||
| GenBank mRNA | JU336583.1 | - | 171 | 104 | ||
| JU477395.1 | - | 171 | 104 | |||
| JV048374.1 | - | 167 | 104 | |||
| JV522067.1 | - | 119 | 104 | |||
More Information
Set Membership: |
|

UBL4A
Genomic Map