Assay Details
| Gene Symbol: TXNIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257935.1 | NP_001244864.1 | 877 | 72 | ||
| GenBank mRNA | JU321896.1 | - | 877 | 72 | ||
| JU470867.1 | - | 894 | 72 | |||
| JV043897.1 | - | 887 | 72 | |||
| JV636533.1 | - | 911 | 72 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TXNIP
Genomic Map