Assay Details
| Gene Symbol: CHEK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149983.2 | XP_015005469.1 | 1190 | 124 | ||
| GenBank mRNA | JU479446.1 | - | 49 | 124 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHEK2| Gene Symbol: CHEK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149983.2 | XP_015005469.1 | 1190 | 124 | ||
| GenBank mRNA | JU479446.1 | - | 49 | 124 | ||
Set Membership: |
|