Assay Details
| Gene Symbol: SAP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266332.1 | NP_001253261.1 | 666 | 117 | ||
| GenBank mRNA | JU326394.1 | - | 377 | 117 | ||
| JU473121.1 | - | 666 | 117 | |||
More Information
Set Membership: |
|

SAP30
Genomic Map