Assay Details
| Gene Symbol: CAMK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028849644.1 | XP_028705477.1 | 873 | 68 | ||
| XM_028849645.1 | XP_028705478.1 | 471 | 68 | |||
| XM_028849646.1 | XP_028705479.1 | 548 | 68 | |||
| XM_028849647.1 | XP_028705480.1 | 547 | 68 | |||
| GenBank mRNA | JU321137.1 | - | 616 | 68 | ||
| JU473372.1 | - | 557 | 68 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028849646.1, XM_028849647.1 transcripts.
Set Membership: |
|

CAMK4
Genomic Map