Assay Details
Gene Symbol: REXO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193813.1 | NP_001180742.1 | 285 | 78 | ||
XM_015115701.1 | XP_014971187.1 | 280 | 78 | |||
XM_028832516.1 | XP_028688349.1 | 236 | 78 | |||
GenBank mRNA | JU333895.1 | - | 170 | 78 | ||
JU475079.1 | - | 165 | 78 | |||
JV046697.1 | - | 182 | 78 | |||
JV647786.1 | - | 162 | 78 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
