Assay Details
| Gene Symbol: CNKSR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265835.1 | NP_001252764.1 | 708 | 84 | ||
| XM_015137078.2 | XP_014992564.2 | 267 | 84 | |||
| XM_028846723.1 | XP_028702556.1 | 249 | 84 | |||
| GenBank mRNA | JU333135.1 | - | 708 | 84 | ||
| JV046284.1 | - | 342 | 84 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015137078.2 transcripts.
Set Membership: |
|

CNKSR3
Genomic Map