Assay Details
| Gene Symbol: FLVCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107314.4 | XP_001107314.2 | 1010 | 90 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLVCR1| Gene Symbol: FLVCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107314.4 | XP_001107314.2 | 1010 | 90 | ||
Set Membership: |
|