Assay Details
| Gene Symbol: NUAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119232.2 | XP_014974718.1 | 508 | 63 | ||
| GenBank mRNA | JV662630.1 | - | 347 | 63 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NUAK2| Gene Symbol: NUAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119232.2 | XP_014974718.1 | 508 | 63 | ||
| GenBank mRNA | JV662630.1 | - | 347 | 63 | ||
Set Membership: |
|