Assay Details
| Gene Symbol: CBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258147.1 | NP_001245076.1 | 2259 | 62 | ||
| XM_028832621.1 | XP_028688454.1 | 1734 | 62 | |||
| GenBank mRNA | JU477627.1 | - | 2259 | 62 | ||
| JU477628.1 | - | 2259 | 62 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CBL
Genomic Map