Assay Details
| Gene Symbol: HEY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194380.3 | NP_001181309.1 | 465 | 90 | ||
| XM_015137303.2 | XP_014992789.1 | 595 | 90 | |||
| XM_015137304.2 | XP_014992790.1 | 624 | 90 | |||
| GenBank mRNA | JU322143.1 | - | 283 | 90 | ||
| JV043955.1 | - | 335 | 90 | |||
| JV637330.1 | - | 341 | 90 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HEY2
Genomic Map