Assay Details
| Gene Symbol: ZFAND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145617.2 | XP_015001103.2 | 312 | 117 | ||
| XM_015145618.2 | XP_015001104.2 | 352 | 117 | |||
| XM_015145619.2 | XP_015001105.2 | 258 | 117 | |||
| GenBank mRNA | JU326031.1 | - | 213 | 117 | ||
| JU473222.1 | - | 297 | 117 | |||
| JV723541.1 | - | 213 | 117 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_015145619.2 transcripts.
This assay is in the 5'UTR region of the XM_015145619.2 transcripts.
Set Membership: |
|

ZFAND1
Genomic Map