Assay Details
| Gene Symbol: ACADSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261772.1 | NP_001248701.1 | 1320 | 92 | ||
| XM_028853439.1 | XP_028709272.1 | 1108 | 92 | |||
| GenBank mRNA | JU334362.1 | - | 1283 | 92 | ||
| JU334363.1 | - | 1283 | 92 | |||
| JU334364.1 | - | 1283 | 92 | |||
| JU334365.1 | - | 1283 | 92 | |||
| JU334366.1 | - | 1283 | 92 | |||
| JU334367.1 | - | 1283 | 92 | |||
| JV046975.1 | - | 1320 | 92 | |||
| JV046976.1 | - | 1320 | 92 | |||
| JV650480.1 | - | 1338 | 92 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ACADSB
Genomic Map