Assay Details
| Gene Symbol: POP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266561.2 | NP_001253490.1 | 419 | 69 | ||
| XM_015152917.2 | XP_015008403.1 | 350 | 69 | |||
| GenBank mRNA | JU334870.1 | - | 437 | 69 | ||
| JU475891.1 | - | 421 | 69 | |||
| JV047292.1 | - | 419 | 69 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

POP5
Genomic Map