Assay Details
| Gene Symbol: HTRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258176.1 | NP_001245105.1 | 1242 | 85 | ||
| GenBank mRNA | JU335497.1 | - | 1242 | 85 | ||
| JV622526.1 | - | 1313 | 85 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HTRA1
Genomic Map