Assay Details
Gene Symbol: H2AFY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261740.1 | NP_001248669.1 | 1134 | 84 | ||
XM_015140865.2 | XP_014996351.1 | 1990 | 84 | |||
XM_015140867.2 | XP_014996353.1 | 1981 | 84 | |||
XM_015140868.2 | XP_014996354.1 | 1978 | 84 | |||
GenBank mRNA | JU337222.1 | - | 997 | 84 | ||
JU337223.1 | - | 1006 | 84 | |||
JU470988.1 | - | 1176 | 84 | |||
JU470989.1 | - | 1170 | 84 | |||
JU477936.1 | - | 1161 | 84 | |||
JV044585.1 | - | 1134 | 84 | |||
JV048777.1 | - | 1125 | 84 | |||
JV600196.1 | - | 1094 | 84 | |||
JV600197.1 | - | 1085 | 84 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
