Assay Details
| Gene Symbol: RBM23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266374.1 | NP_001253303.1 | 1108 | 58 | ||
| XM_015143281.2 | XP_014998767.2 | 1149 | 58 | |||
| XM_015143282.2 | XP_014998768.2 | 1101 | 58 | |||
| XM_015143283.2 | XP_014998769.2 | 1101 | 58 | |||
| XM_015143284.2 | XP_014998770.2 | 905 | 58 | |||
| GenBank mRNA | JU335242.1 | - | 1048 | 58 | ||
| JU335243.1 | - | 1096 | 58 | |||
| JU476212.1 | - | 1076 | 58 | |||
| JU476213.1 | - | 1124 | 58 | |||
| JV047508.1 | - | 1060 | 58 | |||
| JV047509.1 | - | 1108 | 58 | |||
| JV527806.1 | - | 1037 | 58 | |||
| JV527807.1 | - | 1085 | 58 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RBM23
Genomic Map