Assay Details
| Gene Symbol: CBFA2T3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090215.4 | XP_001090215.2 | 1598 | 64 | ||
| XM_002802592.3 | XP_002802638.2 | 1522 | 64 | |||
| XM_015126797.2 | XP_014982283.1 | 1082 | 64 | |||
| XM_028841405.1 | XP_028697238.1 | 1350 | 64 | |||
| XM_028841406.1 | XP_028697239.1 | 1275 | 64 | |||
More Information
Set Membership: |
|

CBFA2T3
Genomic Map