Assay Details
| Gene Symbol: CNOT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267021.1 | NP_001253950.1 | 1103 | 82 | ||
| XM_015141586.2 | XP_014997072.1 | 987 | 82 | |||
| GenBank mRNA | JU328286.1 | - | 432 | 82 | ||
| JU471259.1 | - | 1064 | 82 | |||
| JU476536.1 | - | 1049 | 82 | |||
| JV045480.1 | - | 1070 | 82 | |||
| JV635474.1 | - | 1103 | 82 | |||
More Information
Set Membership: |
|

CNOT6
Genomic Map