Assay Details
| Gene Symbol: FOLH1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115393.2 | XP_014970879.1 | 1171 | 115 | ||
| XM_015115395.2 | XP_014970881.1 | 1171 | 115 | |||
| XM_028833900.1 | XP_028689733.1 | 813 | 115 | |||
| GenBank mRNA | JU341133.1 | - | 1233 | 115 | ||
| JU341134.1 | - | 1233 | 115 | |||
| JV565045.1 | - | 1071 | 115 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FOLH1B
Genomic Map