Assay Details
| Gene Symbol: NEFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144993.2 | XP_015000479.2 | 3596 | 91 | ||
| GenBank mRNA | JV582772.1 | - | 1231 | 91 | ||
| JV617637.1 | - | 1242 | 91 | |||
More Information
Set Membership: |
|

NEFM
Genomic Map