Assay Details
| Gene Symbol: DNAJA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107532.4 | XP_001107532.3 | 614 | 100 | ||
| XM_015142829.2 | XP_014998315.2 | 324 | 100 | |||
| GenBank mRNA | JU321119.1 | - | 390 | 100 | ||
| JV048951.1 | - | 424 | 100 | |||
| JV637749.1 | - | 432 | 100 | |||
More Information
Set Membership: |
|

DNAJA4
Genomic Map