Assay Details
| Gene Symbol: MINDY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133770.2 | XP_014989256.2 | 1932 | 63 | ||
| XM_015133771.2 | XP_014989257.2 | 1887 | 63 | |||
| GenBank mRNA | JV724933.1 | - | 1711 | 63 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MINDY4
Genomic Map