Assay Details
| Gene Symbol: GNAZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261596.1 | NP_001248525.1 | 1257 | 64 | ||
| XM_015149763.2 | XP_015005249.1 | 1306 | 64 | |||
| XM_028827299.1 | XP_028683132.1 | 1354 | 64 | |||
| GenBank mRNA | JU334060.1 | - | 1257 | 64 | ||
| JV680819.1 | - | 1268 | 64 | |||
More Information
Set Membership: |
|

GNAZ
Genomic Map