Assay Details
| Gene Symbol: ASPHD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149964.2 | XP_015005450.1 | 1330 | 74 | ||
| GenBank mRNA | JU335587.1 | - | 913 | 74 | ||
| JU473560.1 | - | 1051 | 74 | |||
| JV045183.1 | - | 1203 | 74 | |||
| JV565730.1 | - | 1277 | 74 | |||
| JV704051.1 | - | 1198 | 74 | |||
More Information
Set Membership: |
|

ASPHD2
Genomic Map