Assay Details
| Gene Symbol: FMO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042777.1 | NP_001036242.1 | 1311 | 88 | ||
| XM_015123252.2 | XP_014978738.1 | 1403 | 88 | |||
| GenBank mRNA | U59453.1 | - | 1311 | 88 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FMO2
Genomic Map